Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics Cobas™ Liat CT/NG RT-PCR Assay Kit

Catalog No. 21200312 Shop All Roche Diagnostics Products
Change view
Click to view available options
Clia Complexity:
Waived
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Clia Complexity
21-200-312 Waived
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-200-312 Supplier Roche Diagnostics Supplier No. 10030933190
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

The Cobas™ liat CT/NG/MG test is an automated, qualitative in vitro nucleic acid diagnostic using real-time PCR for the detection of CT and NG in male urine and vaginal swabs.

The cobas™ liat CT/NG nucleic acid test is an automated, qualitative in vitro nucleic acid diagnostic test that utilizes real- time polymerase chain reaction (PCR) for the direct detection of Chlamydia trachomatis (CT) and Neisseria gonorrhoeae (NG) nucleic acid in male urine and vaginal swabs, all in cobas™ PCR Media (Roche Molecular Systems, Inc.). This test is intended as an aid in the diagnosis of urogenital infections in both symptomatic and asymptomatic individuals.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 20 tests per kit; store at 2°to 8°C
Clia Complexity Waived
Detectable Analytes C. trachomatis, N. gonorrhoeae
Sensitivity Chlamydia: male urine (97.3%); vaginal swab (98.2%). Gonorrhea: male urine (100.0%); vaginal swab (97.7%).
Type PCR test
For Use With (Application) The cobas™ liat CT/NG test is an automated, qualitative in vitro nucleic acid diagnostic using real-time PCR for the detection of CT and NG in male urine and vaginal swabs.
Includes 20 tests per kit; pipettes
Quantity 20 tests per kit
Specificity Chlamydia: male urine (99.9%); vaginal swab (99.8%). Gonorrhea: male urine (99.9%); vaginal swab (99.8%).
Analysis Time 20-minutes
Dimensions (L x W x H) 10.7 x 6.5 x 5.7 in.
Weight (English) 2.3 lb.
Form Ready-to-Use (RTU)
Operating Temperature Range 2° to 8°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.