Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics cobas Liat Strep A Assay

Catalog No. 23-312-199 Shop All Roche Diagnostics Products
Click to view available options
Quantity:
20 Tests/Cs. (Kit)
This item is not returnable. View return policy
Order eligibility is based upon executed contractual agreement.
This item is not returnable. View return policy
Order eligibility is based upon executed contractual agreement.

Qualitative in vitro diagnostic test for detection of Streptococcus pyogenes in throat swab specimens from patients with signs and symptoms of pharyngitis. The Strep A assay utilizes nucleic acid purification and PCR technology to detect Streptococcus pyogenes.

  • Testing accuracy with RT-PCR technology
  • Definitive results with no need for interpretation
  • No confirmation required for negative results
  • Fast, easy operation
  • Three easy steps
  • Results in 15 minutes
TRUSTED_SUSTAINABILITY

Specifications

Accuracy Sensitivity: 98.3%; Specificity: 94.2%
Certifications/Compliance None
Content And Storage 36°F to 46°F
Control Sets Controls purchased separately (FCAT 23312202)
CPT Code 87651
CE Marker No
Clia Complexity Waived
Detectable Analytes S. pyogenes
Format Case (Kit)
Sample Type Throat Swab
Sensitivity 98.3%
Type Nucleic acid RT-PCR Streptococcus pyogenes (Group A) Testing
Packaging Type Case (kit)
Quantity 20 Tests/Cs. (Kit)
Specificity 0.942
Test Time 15 min.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.