Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics cobas™ Liat C. Diff Assay Kit

Catalog No. 23312209
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

The cobas™ Liat C. difficile nucleic acid test for use on the cobas™ Liat System is a real-time PCR test that provides accurate results in 20 minutes for the detection of toxigenic C. diff in unformed stool stamples.

  • The cobas™ Liat C. difficile nucleic acid test for use on the cobas™ Liat System is a real-time PCR test that provides accurate results in 20 minutes for the detection of toxigenic C. diff in unformed stool stamples
  • Provide clinician confidence with results in 20 minutes
  • Reduce enteric isolation time
  • Improves antibiotic stewardship initiatives by de-escalating days of unnecessary antibiotics
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
CPT Code 87493
Description Roche Diagnostics cobas™ Liat C.diff assay kit
CE Marker Yes
Clia Complexity Moderate
Detection Method Target region is C. difficile Toxin B
DoA Calibrators No
Format Kit
Sample Type Unformed stool in cobas™ PCR media
Sensitivity 0.935
Type Assay Test Kit
Disposable Single-use
For Use With (Application) RT-PCR
For Use With (Equipment) Cobas Liat Analyzer
No. of Reactions 20 Cobas Liat C.diff Tests
Packaging Type Kit
Quantity 1 Kit
Specificity 0.964
Test Time 20 min.
Analysis Time 20 min.
Dimensions (L x W x H) 10.75 x 5.83 x 6.7 in.
Weight (English) 2.6 lb.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.