Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COBL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553941
Description
Immunogen sequence: AVVRVSPEVP LQNILPVICA KCEVSPEHVV LLRDNIAGEE LELSKSLNEL GIKELYAWDN RRETFRKSSL GNDETDKEKK KFLGFFKVNK RSNSKGCLT Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.
This gene encodes a cytosolic leucine-tRNA synthetase, a member of the class I aminoacyl-tRNA synthetase family. The encoded enzyme catalyzes the ATP-dependent ligation of L-leucine to tRNA(Leu). It is found in the cytoplasm as part of a multisynthetase complex and interacts with the arginine tRNA synthetase through its C-terminal domain. Alternatively spliced transcript variants of this gene have been found; however, their full-length nature is not known.
Specifications
COBL | |
Polyclonal | |
Unconjugated | |
COBL | |
COBL; cordon-bleu homolog; cordon-bleu WH2 repeat; cordon-bleu WH2 repeat protein; KIAA0633; Protein cordon-bleu | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23242 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75128 | |
COBL | |
Recombinant protein corresponding to Human COBL. Recombinant protein control fragment (Product #RP-92660). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction