Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COBLL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562782
Description
Immunogen sequence: DEKEELSEVP KVEAENISPK SQDIPFVSTD IINTLKNDPD SALGNGSGEF SQNSMEEKQE TKSTDGQEPH SVVYDTSNGK KVVDSIRNLK SLG Highest antigen sequence identity to the following orthologs: Mouse - 52%, Rat - 45%.
The function remains known.
Specifications
COBLL1 | |
Polyclonal | |
Unconjugated | |
COBLL1 | |
1810047P18Rik; Cobll1; COBL-like 1; Coblr1; cobl-related 1; cobl-related protein 1; Cordon-bleu protein-like 1; Cordon-bleu protein-like 1-like protein; cordon-bleu WH2 repeat protein like 1; cordon-bleu WH2 repeat protein-like 1; D430044D16Rik; KIAA0977 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22837 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q53SF7 | |
COBLL1 | |
Recombinant protein corresponding to Human COBLL1. Recombinant protein control fragment (Product #RP-103153). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction