Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COBLL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542173
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542173 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542173 Supplier Invitrogen™ Supplier No. PA542173
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SAPATPLVNK HRPTFTRSNT ISKPYISNTL PSDAPKKRRA PLPPMPASQS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

The function remains known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COBLL1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene COBLL1
Gene Accession No. Q53SF7
Gene Alias 1810047P18Rik; Cobll1; COBL-like 1; Coblr1; cobl-related 1; cobl-related protein 1; Cordon-bleu protein-like 1; Cordon-bleu protein-like 1-like protein; cordon-bleu WH2 repeat protein like 1; cordon-bleu WH2 repeat protein-like 1; D430044D16Rik; KIAA0977
Gene Symbols COBLL1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human COBLL1 (aa 256-305).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22837
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.