Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COBRA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110023
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110023 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110023 Supplier Invitrogen™ Supplier No. PA5110023
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSMILCDPFA INTLALSTVR HLQELVGQET LPRDSPDLLL LLRLLALGQG AWDMIDSQVF KEPKMEVELI TRFLPMLMSF LVDDYTFNVD QKLPAEEKAP VSYPNTLPES FTKFLQEQRM ACEVGLYYVL HITKQRNKNA LLRLLPGLVE TFGDLAFGDI FLHLLTGNLA LLADEFALED FCSSLFDGFF LTASPRKENV HRHALRLLIH LHPRVAPSKL EALQKALEPT GQSGEAVKEL YSQLGEKLEQ LDHRKPSPAQ AAETPALELP LPSVPAPAPL.

NELFB is a subunit of negative elongation factor (NELF), which also includes NELFA (WHSC2), either NELFC or NELFD (TH1L), and NELFE (RDBP), a heterodimer of SPT4 (SUPT4H1), to cause transcriptional pausing of RNA polymerase II (Narita et al., 2003).
TRUSTED_SUSTAINABILITY

Specifications

Antigen COBRA1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NELFB
Gene Accession No. Q8C4Y3, Q8WX92
Gene Alias A730008L03Rik; AB041607; AI663983; Cobra1; cofactor of BRCA1; DKFZp586B0519; KIAA1182; MNCb-5210; Negative elongation factor B; negative elongation factor complex member B; negative elongation factor complex member B, Cobra1; negative elongation factor protein B; NELFB; NELF-B; ortholog of human cofactor of BRCA1 (COBRA1); RGD1307832; RP13-122B23.3
Gene Symbols NELFB
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 349-628 of human NELFB (NP_056271.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25920, 58202
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.