Learn More
COBRA1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301912AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
NELFB is a subunit of negative elongation factor (NELF), which also includes NELFA (WHSC2; MIM 606026), either NELFC or NELFD (TH1L; MIM 605297), and NELFE (RDBP; MIM 154040). NELF acts with DRB sensitivity-inducing factor (DSIF), a heterodimer of SPT4 (SUPT4H1; MIM 603555) and SPT5 (SUPT5H; MIM 602102), to cause transcriptional pausing of RNA polymerase II (see MIM 180660) (Narita et al., 2003 [PubMed 12612062]).Specifications
COBRA1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8WX92 | |
NELFB | |
Between 530 and 580 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
NELFB | |
RP13-122B23.3, DKFZp586B0519, KIAA1182, NELF-B, NELFB, negative elongation factor B; negative elongation factor protein B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25920 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.