Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COCH Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA548475

Catalog No. PA548475


Only null left
Add to Cart

Description

Description

Predicted to react with bovine based on sequence homology.

Cochlin is a secreted protein encoded by the coagulation factor C homology (COCH) gene, a cochlear gene. It constitutes 70% of the inner ear proteins and is classified into three glycosylated isoforms: p63s, p44s and p40. Cochlin contains an N-terminal LCCL domain and two von Willebrand factor A-like domains. Mutations in the COCH gene cause DFNA9, an autosomal dominant nonsyndromic auditory and vestibular dysfunction disorder, as a result of either an amino acid deletion in the LCCL domain or missense substitutions. Microfibrillar deposits accumulate in the inner ear of individuals with DFNA9 and these deposits may contain the Cochlin protein. Cochlin is a target antigen for autoimmune sensorineural hearing loss.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

COCH
Polyclonal
Unconjugated
COCH
AW122937; coagulation factor C homolog (Limulus polyphemus); coagulation factor C homolog, cochlin (Limulus polyphemus); Coch; COCH5B2; COCH-5B2; Cochlin; D12H14S564E; DFNA9; PRO294; UNQ257; UNQ257/PRO294
Rabbit
Antigen affinity chromatography, Protein A
RUO
1690
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.48 mg/mL
PBS with 0.09% sodium azide; pH 7.4
O43405
COCH
KLH conjugated synthetic peptide between 399-428 amino acids from the Central region of human COCH.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.