Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COCH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597602
Description
COCH Polyclonal Antibody for Western Blot, ELISA
Cochlin is a secreted protein encoded by the coagulation factor C homology (COCH) gene, a cochlear gene. It constitutes 70% of the inner ear proteins and is classified into three glycosylated isoforms: p63s, p44s and p40. Cochlin contains an N-terminal LCCL domain and two von Willebrand factor A-like domains. Mutations in the COCH gene cause DFNA9, an autosomal dominant nonsyndromic auditory and vestibular dysfunction disorder, as a result of either an amino acid deletion in the LCCL domain or missense substitutions. Microfibrillar deposits accumulate in the inner ear of individuals with DFNA9 and these deposits may contain the Cochlin protein. Cochlin is a target antigen for autoimmune sensorineural hearing loss.
Specifications
COCH | |
Polyclonal | |
Unconjugated | |
COCH | |
AW122937; coagulation factor C homolog (Limulus polyphemus); coagulation factor C homolog, cochlin (Limulus polyphemus); Coch; COCH5B2; COCH-5B2; Cochlin; D12H14S564E; DFNA9; PRO294; UNQ257; UNQ257/PRO294 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1690 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43405 | |
COCH | |
Recombinant Human Cochlin protein (20-260AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction