Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CoCoA Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP321437C
Description
CoCoA Polyclonal antibody specifically detects CoCoA in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
CoCoA | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
calcium binding and coiled-coil domain 1, calphoglin, Cocoa, Coiled-coil coactivator protein, coiled-coil leucine zipper coactivator 1, inorganic pyrophosphatase activator, KIAA1536calcium-binding and coiled-coil domain-containing protein 1, Sarcoma antigen NY-SAR-3 | |
Produced in rabbits immunized with E. coli-derived Human CoCoA fragment | |
0.1 mL | |
Neuroscience, Signal Transduction | |
57658 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction