Learn More
CoCoA Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
| Antigen | CoCoA |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-155-8527
|
Bethyl Laboratories, Inc
A300818AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat and Orangutan
Functions as a coactivator for aryl hydrocarbon and nuclear receptors (NR). Recruited to promoters through its contact with the N-terminal basic helix-loop-helix-Per-Arnt-Sim (PAS) domain of transcription factors or coactivators, such as NCOA2. During ER-activation acts synergistically in combination with other NCOA2-binding proteins, such as EP300, CREBBP and CARM1. Involved in the transcriptional activation of target genes in the Wnt/CTNNB1 pathway. Functions as a secondary coactivator in LEF1-mediated transcriptional activation via its interaction with CTNNB1. Coactivator function for nuclear receptors and LEF1/CTNNB1 involves differential utilization of two different activation regions. Seems to enhance inorganic pyrphosphatase thus activating phosphogluomutase (PMG). Probably functions as component of the calphoglin complex, which is involved in linking cellular metabolism (phosphate and glucose metabolism) with other core functions including protein synthesis and degradation, calcium signaling and cell growth.Specifications
| CoCoA | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Cocoa, calphoglin, calcium-binding and coiled-coil domain-containing protein 1, coiled-coil coactivator protein, coiled-coil leucine zipper coactivator 1, inorganic pyrophosphatase activator, sarcoma antigen NY-SAR-3 | |
| Between 650 and C-term | |
| Primary | |
| 4° C | |
| CALCOCO1 |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q8CGU1, Q9P1Z2 | |
| 57658, 67488 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.