Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Coconut oil, pure, refined

Catalog No. p-2691232
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 8001-31-8

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8001-31-8
MDL Number MFCD00130817

Specifications

Acidity 0.5mg KOH/g max.
Melting Point 23.0°C to 26.0°C
Color White
Density 0.9030g/mL
Flash Point >200°C
Packaging Glass bottle
Quantity 100 g
Specific Gravity 0.903
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,chloroform and carbon,disulfide
Grade Pure
Physical Form Unctious Mass
Chemical Name or Material Coconut oil, Refined
Show More Show Less
EINECSNumber 232-282-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.