Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ SMART™ Spectro System COD Reagents
SDPSCI_ED

Catalog No. s72412 Shop All LaMotte Company Products
Click to view available options
Range:
0 to 15,000ppm
0 to 1500ppm
0 to 150ppm

A precise and easy way to measure critical COD levels.

Measure low, medium or high levels of Chemical Oxygen Demand (COD) using a mercury reagent system approved by the Environmental Protection Agency (EPA).

  • Comes in 25 vials to a case
  • Requires COD Heater Block, not included

Specifications

Certifications/Compliance EPA
For Use With (Equipment) LaMotte™ colorimeters
Range 0 to 150ppm
Test Method Acid digestion
Length (English) 0.62 in.
Length (Metric) 1.6 cm
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.