Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Code Breakers: Using CRISPR to Rewrite Genetics
SDPSCI_ED

Catalog No. S05963 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S05963 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05963 Supplier EDVOTEK™ Supplier No. 307
Add to Cart
Add to Cart

In this 'Code Breakers' experiment, students explore cutting-edge genetic engineering using CRISPR-Cas9 to knock out the purple chromogenic genes in classroom safe bacteria. Additionally, students test the specificity of CRISPR-Cas9 by using a non-specific CRISPR gRNA template.

  • Explore genetic engineering, CRISPR, and the central dogma of molecular biology in this comprehensive experiment.
  • Transform classroom- safeE. coli with CRISPR RNA templates to test the targeting specificity of the technology.
  • Identify successful gene knockout based on the color of the cells.
  • Group Size: For 10 Lab Groups
  • Time Required: Set Up - 50 minutes / Incubation - overnight / Transformation - 90 minutes
  • Kit Includes: Instructions, LyphoCells™E.coli Purple, ampicillin, IPTG, Chloramphenicol, ReadyPour™ SOC-agar, pCas9-PurpleGuide (plasmid), pCas9-ControlGuide (plasmid), CaCl2, competent cell solution, SOC liquid medium (broth), SOC additive (for ReadyPour ONLY), petri plates (small), petri plates (large), plastic microtipped transfer pipets, wrapped 10 ml pipet (sterile), inoculating loops (sterile), microcentrifuge tubes, 50mL conical tubes
  • All You Need:Automatic micropipette (5-50 μl) and tips, two water baths (37°C and 42°C), incubation oven (optional but recommended), thermometer, centrifuge(s), vortexer (optional), floating racks, pipet pumps or bulbs, ice, marking pens, tape, hot plate or microwave oven, and hot gloves
  • Storage: Some Components Require Freezer Storage Upon Receipt.

Specifications

Grade 9 to Undergraduate
Class Size 10 Lab Groups
Product Type Biotechnology Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.