Learn More
Brady™ Code Reader™ 2600 Barcode Readers
Catalog No. 19104470 Shop All Brady Corporation Products
Change view
Click to view available options
Includes:
Bluetooth, Battery
Bluetooth, Battery, Charging Station, M3 Modem with USB Comm Cable
Type:
Barcode Reader Handle Kit
Barcode Reader Palm Kit
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Type
19-104-470 Bluetooth, Battery Barcode Reader Handle Kit
19-104-469 Bluetooth, Battery, Charging Station, M3 Modem with USB Comm Cable Barcode Reader Handle Kit
19-104-471 Bluetooth, Battery, Charging Station, M3 Modem with USB Comm Cable Barcode Reader Palm Kit
19-104-472 Bluetooth, Battery Barcode Reader Palm Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-104-470 Supplier Brady™ Supplier No. 133081
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Reads all 1D and 2D barcode symbologies
Reads all 1D and 2D barcode symbologies
  • Easy to use, lightweight, and high performing
  • Features durable, rechargeable battery plus programmable buttons for custom workflow
  • Battery status LED indicators with fuel gauge
  • User feedback with vibration, audible tones and LED
  • Patented dual field optics, both high density and wide field in same unit
  • High speed, omnidirectional reading of 1D and 2D barcodes
  • Unequaled performance on rounded, curved and shiny surfaces
  • Multiple programmable buttons for customized work flow processes
  • Easy to clean, disinfectant ready, IP65 housing for healthcare use
  • Powerful data management capability with JavaScript
  • Available in Handle and Palm kits
Specifications
Includes Bluetooth, Battery
Type Barcode Reader Handle Kit