Description
Reads all 1D and 2D barcode symbologies
- Easy to use, lightweight, and high performing
- Features durable, rechargeable battery plus programmable buttons for custom workflow
- Battery status LED indicators with fuel gauge
- User feedback with vibration, audible tones and LED
- Patented dual field optics, both high density and wide field in same unit
- High speed, omnidirectional reading of 1D and 2D barcodes
- Unequaled performance on rounded, curved and shiny surfaces
- Multiple programmable buttons for customized work flow processes
- Easy to clean, disinfectant ready, IP65 housing for healthcare use
- Powerful data management capability with JavaScript
- Available in Handle and Palm kits
Specifications
Specifications
| Includes | Bluetooth, Battery, Charging Station, M3 Modem with USB Comm Cable |
| Type | Barcode Reader Palm Kit |