Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ Coelenterazine

Catalog No. C2944
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2944 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2944 Supplier Molecular Probes™ Supplier No. C2944
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Coelenterazine

Coelenterazine and synthetic coelenterazine analogs are useful for reconstituting aequorin in cells that have been transfected with apoaequorin cDNA.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Bioluminescence
Product Type Coelenterazine
Dye Type Protein-Based
Quantity 250 μg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.