Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cofilin 1/2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115034
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115034 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115034 Supplier Invitrogen™ Supplier No. PA5115034
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Cofilin.

Cofilin is a widely distributed intracellular actin-modulating protein that binds and depolymerizes filamentous F-actin and inhibits the polymerization of monomeric G-actin in a pH-dependent manner. It is involved in the translocation of actin-cofilin complex from cytoplasm to nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cofilin 1/2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene CFL1
Gene Accession No. P18760, P23528, P45591, P45592, Q9Y281
Gene Alias 18 kDa phosphoprotein; AA959946; CFL; CFL1; CFL-1; CFL2; Cof; cofilin; cofilin 1; cofilin 1 (non-muscle); cofilin 1, non-muscle; cofilin 2; cofilin 2 (muscle); cofilin 2, muscle; cofilin, muscle isoform; Cofilin, non-muscle isoform; Cofilin1; cofilin-1; Cofilin-2; epididymis secretory protein Li 15; HEL-S-15; n-cofilin; NEM7; nemaline myopathy type 7; p18
Gene Symbols CFL1, CFL2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CFL1(Accession P23528), corresponding to amino acid residues K114-K164.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1072, 1073, 12631, 12632, 29271, 366624
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.