Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Cofilin-1, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABT1493 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABT1493 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT1493 Supplier MilliporeSigma™ Supplier No. ABT1493
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cofilin-1 (UniProt P23528; also known as 18kDa phosphoprotein, Cofilin non-muscle isoform, p18) is encoded by the CFL1 (also known as CFL) gene (Gene ID 1072) in human. Cofilin-1 (non-muscle cofilin), cofilin-2 (muscle cofilin), and actin-depolymerizing factor (ADF) play an essential role in regulating actin filament dynamics and reorganization by stimulating the severance and depolymerization of actin filaments. The three cofilin family members share 70% to 81% sequence identity, exhibit similar biological functions, and display distinct but overlapping expression patterns. Cofilins and ADF bind actin monomers and filaments (G-actin and F-actin) and increase the number of actin monomers and filament fragments, facilitating filament turnover and treadmilling at key locations in migrating cells. Cofilins and ADF are inactivated by phosphorylation at Ser3 by LIM-kinases (LIMKs) and testicular protein kinases (TESKs) and are reactivated by dephosphorylation by the slingshot (SSH) family of protein phosphatases and chronophin. Cfl1-knockout results in mouse embryonic lethality at E11.5 12.5 due to aberrant neural tube closure and defective neural crest cell migration, while Adf-deficient mice are viable with normal brain appearance, but suffer from corneal defects that cause blindness in adult mice.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cofilin-1
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene CFL, CFL1
Gene Accession No. P23528
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the N-termnal end sequence of human cofilin-1.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_005498
Test Specificity This rabbit polyclonal antibody targets cofilin-1 N-terminal end sequence conserved among bovine, human, mouse, porcine, and rat species.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.