Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cofilin 2 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298909PE
Description
Cofilin 2 Polyclonal antibody specifically detects Cofilin 2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Cofilin 2 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
cofilin 2 (muscle), Cofilin, muscle isoform, cofilin-2, NEM7 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Cofilin 2. | |
0.1 mL | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
1073 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction