Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cofilin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674613
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-613 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-613 Supplier Invitrogen™ Supplier No. PA19029
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

Cofilin is a widely distributed intracellular actin-modulating protein that binds and depolymerizes filamentous F-actin and inhibits the polymerization of monomeric G-actin in a pH-dependent manner. It is involved in the translocation of actin-cofilin complex from cytoplasm to nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cofilin 2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CFL2
Gene Accession No. Q9Y281
Gene Alias CFL2; COF2; cofilin 2; cofilin 2 (muscle); cofilin 2, muscle; cofilin, muscle isoform; Cofilin-2; HGNC:1875; NEM7; nemaline myopathy type 7
Gene Symbols CFL2
Host Species Goat
Immunogen Synthetic peptide corresponding to the C terminus amino acids DIKDRSTLGEK (aa 142-152).
Purification Method Ammonium sulfate precipitation, Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1073, 366624
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.