Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cofilin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | Cofilin |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 |
Applications | Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017828
![]() |
Novus Biologicals
NB100818660.025ML |
0.025 mL |
Each for $181.90
|
|
|||||
NB10081866
![]() |
Novus Biologicals
NB10081866 |
0.1 mL |
Each for $487.50
|
|
|||||
Description
Cofilin Polyclonal specifically detects Cofilin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cofilin | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
Polyclonal | |
Rabbit | |
Cytoskeleton Markers | |
P23528 | |
1072 | |
Peptide sequence around aa. 1∼5 (M-A-S-G-V) derived from Human cofilin. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
18 kDa phosphoprotein, CFL, cofilin 1 (non-muscle), Cofilin, non-muscle isoform, cofilin-1, p18 | |
CFL1 | |
IgG | |
Affinity Purified | |
19 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title