Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cofilin Monoclonal Antibody (C12), Invitrogen™

Mouse Monoclonal Antibody

$195.80 - $426.00

Specifications

Antigen Cofilin
Clone C12
Concentration 1 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FEMA551063
View Documents
Invitrogen™
MA551063
200 μL
Each for $426.00
Only null left
Add to Cart
 
FEMA551064
View Documents
Invitrogen™
MA551064
20 μL
Each for $195.80
Only null left
Add to Cart
 
Description

Description

Cofilin binds to F-actin and exhibits pH-sensitive F-actin depolymerizing activity. Regulates actin cytoskeleton dynamics. Important for normal progress through mitosis and normal cytokinesis. Plays a role in the regulation of cell morphology and cytoskeletal organization. Required for the up-regulation of atypical chemokine receptor ACKR2 from endosomal compartment to cell membrane, increasing its efficiency in chemokine uptake and degradation. Required for neural tube morphogenesis and neural crest cell migration.
Specifications

Specifications

Cofilin
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
18 kDa phosphoprotein; AA959946; CFL; Cfl1; CFL-1; Cof; COF1; cofilin; cofilin 1; cofilin 1 (non-muscle); cofilin 1, non-muscle; cofilin, non-muscle isoform; Cofilin1; Cofilin-1; cofilin-1-like protein; epididymis secretory protein Li 15; HEL-S-15; n-cofilin; p18
CFL1
IgG2a κ
Protein A/G
CFL1
C12
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Unconjugated
Mouse
Human, Bovine, Pig
P10668, P23528, Q5E9F7
1072, 445532, 534553
Recombinant Human CFL1 protein, Ser3-Leu161 (Accession #P23528).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.