Learn More
Invitrogen™ Cofilin Monoclonal Antibody (GT217)

Description
Positive Control: 293T, A431, HeLa, HepG2, PC-12, Rat-2, Neuro2A, C8D30, NIH-3T3, Raw264.7, C2C12 Predicted Reactivity: Pig (99%), Sheep (99%), Rhesus Monkey (99%), Bovine (99%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Cofilin |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | GT217 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and no preservative; pH 7 |
| Gene | CFL1 |
| Gene Accession No. | P18760, P23528, P45592 |
| Gene Alias | 18 kDa phosphoprotein; AA959946; CFL; Cfl1; CFL-1; Cof; COF1; cofilin; cofilin 1; cofilin 1 (non-muscle); cofilin 1, non-muscle; cofilin, non-muscle isoform; Cofilin1; Cofilin-1; cofilin-1-like protein; epididymis secretory protein Li 15; HEL-S-15; n-cofilin; p18 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.