Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cofilin, p Ser3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313302
This item is not returnable.
View return policy
Description
Cofilin Polyclonal antibody specifically detects Cofilin in Human, Mouse samples. It is validated for Western BlotSpecifications
| Cofilin | |
| Polyclonal | |
| PBS, 1% BSA, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| 18 kDa phosphoprotein, CFL, cofilin 1 (non-muscle), Cofilin, non-muscle isoform, cofilin-1, p18 | |
| Carrier-protein conjugated synthetic peptide corresponding to residues around human Cofilin (phospho Ser3). The exact sequence is proprietary. | |
| 100 μg | |
| Cell Biology, Cellular Markers, Neuronal Cell Markers, Phospho Specific | |
| 1072 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction