Learn More
Invitrogen™ Cofilin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595291
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human 293T whole cell, human U-87 MG whole cell, monkey COS-7 whole cell, rat brain tissue, rat C6 whole cell, mouse brain tissue. IHC: mouse intestine tissue, rat intestine tissue, human intestinal cancer tissue. ICC/IF: MCF-7 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Cofilin binds to F-actin and exhibits pH-sensitive F-actin depolymerizing activity. Regulates actin cytoskeleton dynamics. Important for normal progress through mitosis and normal cytokinesis. Plays a role in the regulation of cell morphology and cytoskeletal organization. Required for the up-regulation of atypical chemokine receptor ACKR2 from endosomal compartment to cell membrane, increasing its efficiency in chemokine uptake and degradation. Required for neural tube morphogenesis and neural crest cell migration.
Specifications
Cofilin | |
Polyclonal | |
Unconjugated | |
CFL1 | |
18 kDa phosphoprotein; AA959946; CFL; Cfl1; CFL-1; Cof; COF1; cofilin; cofilin 1; cofilin 1 (non-muscle); cofilin 1, non-muscle; cofilin, non-muscle isoform; Cofilin1; Cofilin-1; cofilin-1-like protein; epididymis secretory protein Li 15; HEL-S-15; n-cofilin; p18 | |
Rabbit | |
Affinity chromatography | |
RUO | |
1072, 12631, 29271 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
P18760, P23528, P45592 | |
CFL1 | |
A synthetic peptide corresponding to a sequence at the C-terminus of human Cofilin (122-152aa DAIKKKLTGIKHELQANCYEEVKDRCTLAEK). | |
100 μg | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.