Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cofilin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595869
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595869 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595869 Supplier Invitrogen™ Supplier No. PA595869
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MASGVAVSDG VIKVFNDMKV RKSSTPEEVK KRKKAVLFCL SEDKKNIILE EGKEILVGDV GQTVDDPYAT FVKMLPDKDC RYALYDATYE TKESKKEDLV; Positive Samples: HeLa, Jurkat, SH-SY5Y, MCF-7, Mouse brain, Mouse testis, Rat brain, Rat ovary; Cellular Location: Cell projection, Cytoplasm, Cytoplasmic side, Nucleus matrix, Peripheral membrane protein, cytoskeleton, lamellipodium membrane, ruffle membrane.

Cofilin binds to F-actin and exhibits pH-sensitive F-actin depolymerizing activity. Regulates actin cytoskeleton dynamics. Important for normal progress through mitosis and normal cytokinesis. Plays a role in the regulation of cell morphology and cytoskeletal organization. Required for the up-regulation of atypical chemokine receptor ACKR2 from endosomal compartment to cell membrane, increasing its efficiency in chemokine uptake and degradation. Required for neural tube morphogenesis and neural crest cell migration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cofilin
Applications ELISA, Western Blot, Western Blot
Classification Polyclonal
Concentration 0.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene CFL1
Gene Accession No. P18760, P23528, P45592
Gene Alias 18 kDa phosphoprotein; AA959946; CFL; Cfl1; CFL-1; Cof; COF1; cofilin; cofilin 1; cofilin 1 (non-muscle); cofilin 1, non-muscle; cofilin, non-muscle isoform; Cofilin1; Cofilin-1; cofilin-1-like protein; epididymis secretory protein Li 15; HEL-S-15; n-cofilin; p18
Gene Symbols CFL1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CFL1 (NP_0054981).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1072, 12631, 29271
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.