Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cofilin Rabbit anti-Human, Mouse, Rat, Clone: S06-2C0, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
Antigen | Cofilin |
---|---|
Clone | S06-2C0 |
Dilution | Western Blot, ELISA, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 5 μg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
Cofilin Monoclonal antibody specifically detects Cofilin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Cofilin | |
Western Blot, ELISA, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 5 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
18 kDa phosphoprotein, CFL, cofilin 1 (non-muscle), Cofilin, non-muscle isoform, cofilin-1, p18 | |
A synthetic peptide of human Cofilin (Uniprot#: P23528) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S06-2C0 | |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cell Biology, Cellular Markers, Neuronal Cell Markers | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
1072 | |
IgG | |
Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title