Learn More
Invitrogen™ COG1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total COG1 protein.
Specifications
Specifications
| Antigen | COG1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 2.7 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | COG1 |
| Gene Accession No. | Q8WTW3 |
| Gene Alias | 6030445I15; CDG2G; COG complex subunit 1; Cog1; Component of oligomeric Golgi complex 1; conserved oligomeric Golgi complex protein 1; conserved oligomeric Golgi complex subunit 1; DKFZp762L1710; KIAA1381; Ldlb; low density lipoprotein B; low density lipoprotein receptor defect B complementing; low density lipoprotein receptor defect B-complementing protein; mKIAA1381 |
| Gene Symbols | COG1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.