Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556085
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556085 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556085 Supplier Invitrogen™ Supplier No. PA556085
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FCSALDSKLK VKLDDLLAYL PSDDSSLPKD VSPTQAKSSA FDRYADAGTV QEMLRTQSVA CIKHIVDCIR AELQSIEEGV QGQQDALNSA KLHSVLFMA Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 76%.

The protein encoded by this gene is one of eight proteins which form a Golgi-localized complex required for normal Golgi morphology and function. It is thought that this protein is required for steps in the normal medial and trans Golgi-associated processing of glycoconjugates and plays a role in the organization of the Golgi-localized complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COG1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COG1
Gene Accession No. Q8WTW3
Gene Alias 6030445I15; CDG2G; COG complex subunit 1; Cog1; Component of oligomeric Golgi complex 1; conserved oligomeric Golgi complex protein 1; conserved oligomeric Golgi complex subunit 1; DKFZp762L1710; KIAA1381; Ldlb; low density lipoprotein B; low density lipoprotein receptor defect B complementing; low density lipoprotein receptor defect B-complementing protein; mKIAA1381
Gene Symbols COG1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COG1. Recombinant protein control fragment (Product #RP-93371).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9382
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.