Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567442
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567442 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567442 Supplier Thermo Scientific Supplier No. PA567442
Add to Cart
Add to Cart

COG2 Polyclonal Antibody

COG2 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen COG2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cog2
Gene Accession No. Q14746
Gene Alias 1190002B08Rik; 2700012E02Rik; brefeldin A-sensitive, peripheral Golgi protein; C86089; COG 2; COG complex subunit 2; COG2; Component of oligomeric Golgi complex 2; conserved oligomeric Golgi complex protein 2; conserved oligomeric Golgi complex subunit 2; LDLC; low density lipoprotein receptor defect C complementing; low density lipoprotein receptor defect C-complementing protein; Transcript ch4822
Gene Symbols Cog2
Host Species Rabbit
Immunogen Recombinant Human COG2. Recombinant protein control fragment (Product #RP-94222).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22796
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.