Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576872
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576872 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576872 Supplier Invitrogen™ Supplier No. PA576872
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

The structure and function of the Golgi apparatus is controlled by a number of multi-protein complexes that are involved in glycosylation reactions and vesicular transport. The conserved oligomeric Golgi (COG) complex consists of three subcomplexes, termed LDLC, SEC34 and GTT (Golgi transport complex), all of which contain proteins necessary for proper Golgi operation. COG2 (conserved oligomeric Golgi complex subunit 2), also known as LDLC, is a 730 amino acid component of the COG complex. Localized to the cytoplasmic side of the Golgi apparatus, COG2 is required for proper Golgi morphology and function, specifically playing a role in Golgi ribbon formation and vesicular transport. Abnormal COG2 function may cause cell death, suggesting that COG2 is an important factor in cell viability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COG2
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Cog2
Gene Accession No. Q14746, Q921L5
Gene Alias 1190002B08Rik; 2700012E02Rik; brefeldin A-sensitive, peripheral Golgi protein; C86089; COG 2; COG complex subunit 2; COG2; Component of oligomeric Golgi complex 2; conserved oligomeric Golgi complex protein 2; conserved oligomeric Golgi complex subunit 2; LDLC; low density lipoprotein receptor defect C complementing; low density lipoprotein receptor defect C-complementing protein; Transcript ch4822
Gene Symbols Cog2
Host Species Rabbit
Immunogen Recombinant full length Human COG2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22796, 690961, 76332
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.