Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COG4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | COG4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
COG4 Polyclonal specifically detects COG4 in Human samples. It is validated for Western Blot.Specifications
COG4 | |
Polyclonal | |
Rabbit | |
Golgi Apparatus Markers | |
CDG2J, COD1complexed with Dor1p, COG complex subunit 4, component of oligomeric golgi complex 4DKFZp586E1519, conserved oligomeric Golgi complex protein 4, conserved oligomeric Golgi complex subunit 4, DKFZP586E1519 | |
COG4 | |
IgG | |
This product is specific to Subunit or Isoform: 4. |
Western Blot | |
Unconjugated | |
RUO | |
Q9H9E3 | |
25839 | |
Synthetic peptides corresponding to COG4(component of oligomeric golgi complex 4) The peptide sequence was selected from the middle region of COG4. Peptide sequence LFSQGIGGEQAQAKFDSCLSDLAAVSNKFRDLLQEGLTELNSTAIKPQVQ. | |
Primary | |
89 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title