Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COG4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559355
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559355 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559355 Supplier Invitrogen™ Supplier No. PA559355
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILDLKFCMDG VQTALRSEDY EQAAAHIHRY LCLDKSVIEL SRQGKEGSMI DANLKLLQEA EQRLKAIVAE KFAIATKEGD LPQVERFFKI FPLL Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

Multiprotein complexes are key determinants of Golgi apparatus structure and its capacity for intracellular transport and glycoprotein modification. Several complexes have been identified, including the Golgi transport complex (GTC), the LDLC complex, which is involved in glycosylation reactions, and the SEC34 complex, which is involved in vesicular transport. These 3 complexes are identical and have been termed the conserved oligomeric Golgi (COG) complex, which includes COG4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COG4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COG4
Gene Accession No. Q9H9E3
Gene Alias AW554810; CDG2J; COD1; COG complex subunit 4; COG4; complexed with Dor1p; component of oligomeric golgi complex 4; conserved oligomeric Golgi complex protein 4; conserved oligomeric Golgi complex subunit 4; D8Ertd515e
Gene Symbols COG4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COG4. Recombinant protein control fragment (Product #RP-98540).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25839
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.