Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COG6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559180
Description
Immunogen sequence: VQQHKPEQGS LANMPNLDSV TLKAAMVQFD RYLSAPDNLL IPQLNFLLSA TVKEQIVKQS TELVCRAYGE VYAAVMNPIN EYKDPENILH RSPQQVQTLL Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.
This gene encodes a subunit of the conserved oligomeric Golgi complex that is required for maintaining normal structure and activity of the Golgi apparatus. The encoded protein is organized with conserved oligomeric Golgi complex components 5, 7 and 8 into a sub-complex referred to as lobe B. Alternative splicing results in multiple transcript variants.
Specifications
COG6 | |
Polyclonal | |
Unconjugated | |
COG6 | |
4933405E16Rik; AU018618; CDG2L; COD2; COG complex subunit 6; COG6; complexed with Dor1p 2; component of oligomeric golgi complex 6; conserved oligomeric Golgi complex protein 6; conserved oligomeric Golgi complex subunit 6; Conserved oligomeric Golgi complex subunit 6-like protein; hypothetical protein MGC56117; KIAA1134; LOW QUALITY PROTEIN: conserved oligomeric Golgi complex subunit 6; mKIAA1134; SHNS; si:ch211-203f4.1; testicular tissue protein Li 41; zgc:56117 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57511 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2V7 | |
COG6 | |
Recombinant protein corresponding to Human COG6. Recombinant protein control fragment (Product #RP-97947). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction