Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COG6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587806
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587806 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587806 Supplier Invitrogen™ Supplier No. PA587806
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKTTLALFEF TDRRLEMLQF QIEAHLDTLI NEQASYVLTR VGLSYIYNTV QQHKPEQGSL ANMPNLDSVT LKAAMVQFDR YLSAPDNLLI PQLNFLLSAT VKEQIVKQST ELVCRAYGEV YAAVMNPINE YKDPENILHR SPQQVQTLLS; Positive Samples: Jurkat, A-549, U-87MG, Mouse kidney, Rat testis; Cellular Location: Golgi apparatus membrane, Peripheral membrane protein.

This gene encodes a subunit of the conserved oligomeric Golgi complex that is required for maintaining normal structure and activity of the Golgi apparatus. The encoded protein is organized with conserved oligomeric Golgi complex components 5, 7 and 8 into a sub-complex referred to as lobe B. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COG6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene COG6
Gene Accession No. Q68FP9, Q8R3I3, Q9Y2V7
Gene Alias 4933405E16Rik; AU018618; CDG2L; COD2; COG complex subunit 6; COG6; complexed with Dor1p 2; component of oligomeric golgi complex 6; conserved oligomeric Golgi complex protein 6; conserved oligomeric Golgi complex subunit 6; Conserved oligomeric Golgi complex subunit 6-like protein; hypothetical protein MGC56117; KIAA1134; LOW QUALITY PROTEIN: conserved oligomeric Golgi complex subunit 6; mKIAA1134; SHNS; si:ch211-203f4.1; testicular tissue protein Li 41; zgc:56117
Gene Symbols COG6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 508-657 of human COG6 (NP_0658021).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310411, 57511, 67542
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.