Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COG7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | COG7 |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Description
COG7 Polyclonal specifically detects COG7 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
COG7 | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
91949 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LPELDNMADNWLGSIARATMQTYCDAILQIPELSPHSAKQLATDIDYLINVMDALGLQPSRTLQHIVTLLKTRPEDYRQVSKGLPRRLATTVATMRSVN | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Polyclonal | |
Rabbit | |
Human | |
CDG2E, component of oligomeric golgi complex 7COG complex subunit 7, conserved oligomeric Golgi complex subunit 7 | |
COG7 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title