Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COG7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559285
Description
Immunogen sequence: ISAKLTGMQN SLMMLVDTPD YSEKCVHLEA LKNRLEALAS PQIVAAFTSQ AVDQSKVFVK VFTEIDRMPQ LLAYYYKCHK VQLLAAWQEL CQSDLSLDRQ Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
This gene encodes a WD repeat-containing protein that interacts with the Cul4-Ddb1 E3 ligase macromolecular complex. Multiple alternatively spliced transcript variants have been found for this gene.
Specifications
COG7 | |
Polyclonal | |
Unconjugated | |
COG7 | |
5630400E24Rik; CDG2E; COG complex subunit 7; Cog7; Component of oligomeric Golgi complex 7; conserved oligomeric Golgi complex subunit 7; Gm167; UNQ3082/PRO10013 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
91949 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P83436 | |
COG7 | |
Recombinant protein corresponding to Human COG7. Recombinant protein control fragment (Product #RP-98582). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction