Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COG7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584660
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584660 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584660 Supplier Invitrogen™ Supplier No. PA584660
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPELDNMADN WLGSIARATM QTYCDAILQI PELSPHSAKQ LATDIDYLIN VMDALGLQPS RTLQHIVTLL KTRPEDYRQV SKGLPRRLAT TVATMRSVN Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 88%.

This gene encodes a WD repeat-containing protein that interacts with the Cul4-Ddb1 E3 ligase macromolecular complex. Multiple alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COG7
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COG7
Gene Accession No. P83436
Gene Alias 5630400E24Rik; CDG2E; COG complex subunit 7; Cog7; Component of oligomeric Golgi complex 7; conserved oligomeric Golgi complex subunit 7; Gm167; UNQ3082/PRO10013
Gene Symbols COG7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COG7. Recombinant protein control fragment (Product #RP-105327).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91949
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.