Learn More
COG7 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305506AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Multiprotein complexes are key determits of Golgi apparatus structure and its capacity for intracellular transport and glycoprotein modification. Several complexes have been identified, including the Golgi transport complex (GTC), the LDLC complex, which is involved in glycosylation reactions, and the SEC34 complex, which is involved in vesicular transport. These 3 complexes are identical and have been termed the conserved oligomeric Golgi (COG) complex, which includes COG7 (Ungar et al., 2002 [PubMed 11980916]).Specifications
COG7 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P83436 | |
COG7 | |
Between 720 and 770 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
COG7 | |
COG7; component of oligomeric golgi complex 7; CDG2E | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91949 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.