Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COG7 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519031
Description
This antibody is predicted to react with canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
This gene encodes a WD repeat-containing protein that interacts with the Cul4-Ddb1 E3 ligase macromolecular complex. Multiple alternatively spliced transcript variants have been found for this gene.
Specifications
COG7 | |
Polyclonal | |
Unconjugated | |
COG7 | |
5630400E24Rik; CDG2E; COG complex subunit 7; Cog7; Component of oligomeric Golgi complex 7; conserved oligomeric Golgi complex subunit 7; Gm167; UNQ3082/PRO10013 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
91949 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P83436 | |
COG7 | |
Synthetic peptide sequence (KTRPEDYRQVSK) corresponding to the internal amino acids of COG7 (aa 741-753). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction