Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COG8 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP215938

Catalog No. NBP215938


Only null left
Add to Cart

Description

Description

COG8 Polyclonal specifically detects COG8 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

COG8
Polyclonal
Unconjugated
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
COG complex subunit 8, component of oligomeric golgi complex 8CDG2H, conserved oligomeric Golgi complex subunit 8, dependent on RIC1, DOR1conserved oligomeric golgi complex component 8, FLJ22315
Rabbit
68 kDa
0.1 mL
Primary
Human. Expected to cross react based on sequence identity: Mouse (89%), Rat (91%), Bovine (90%).
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Western Blot, Immunocytochemistry, Immunofluorescence
1mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Q96MW5
COG8
Recombinant protein encompassing a sequence within the center region of human COG8. The exact sequence is proprietary.
Affinity Purified
RUO
84342
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only