Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565371
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565371 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565371 Supplier Invitrogen™ Supplier No. PA565371
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSVIKIHTLVE SLKLSITDQQ LPMFIRIMQL GIALYYGEIG NFKEGEIEDL TCHNKDMLGN ITGSEDETR Highest antigen sequence identity to the following orthologs - mouse 83%, rat 27%.

COH1 (Cohen syndrome protein 1), also known as VPS13B (vacuolar protein sorting-associated protein 13B) or CHS1, belongs to the VPS13 family and may function in vesicle-mediated transport and sorting of proteins within the cell. COH1 is widely expressed and multiple alternatively spliced transcript variants have been observed. Mutations in this gene have been associated with Cohen syndrome. COH1 is a Golgi-localized peripheral membrane protein and plays a critical role in Golgi (re)assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COH1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VPS13B
Gene Accession No. Q7Z7G8
Gene Alias 1810042B05Rik; 2310042E16Rik; 4732488H20; C330002D13Rik; C87206; CHS1; COH1; Cohen syndrome homolog 1; Cohen syndrome protein 1; Cohen syndrome protein 1 homolog; D230005K13; KIAA0532; mKIAA0532; mKIAA5032; vacuolar protein sorting 13 homolog B; vacuolar protein sorting 13B (yeast); vacuolar protein sorting-associated protein 13B; Vps13b
Gene Symbols VPS13B
Host Species Rabbit
Immunogen Recombinant Human COH1. Recombinant protein control fragment (Product #RP-104778).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157680
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.