Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534406
Description
A suggested positive control is SK-N-SH cell lysate. PA5-34406 can be used with blocking peptide PEP-1448.
COH1 (Cohen syndrome protein 1), also known as VPS13B (vacuolar protein sorting-associated protein 13B) or CHS1, belongs to the VPS13 family and may function in vesicle-mediated transport and sorting of proteins within the cell. COH1 is widely expressed and multiple alternatively spliced transcript variants have been observed. Mutations in this gene have been associated with Cohen syndrome. COH1 is a Golgi-localized peripheral membrane protein and plays a critical role in Golgi (re)assembly.
Specifications
| COH1 | |
| Polyclonal | |
| Unconjugated | |
| VPS13B | |
| 1810042B05Rik; 2310042E16Rik; 4732488H20; C330002D13Rik; C87206; CHS1; COH1; Cohen syndrome homolog 1; Cohen syndrome protein 1; Cohen syndrome protein 1 homolog; D230005K13; KIAA0532; mKIAA0532; mKIAA5032; vacuolar protein sorting 13 homolog B; vacuolar protein sorting 13B (yeast); vacuolar protein sorting-associated protein 13B; Vps13b | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 157680, 315036, 666173 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q7Z7G8, Q80TY5 | |
| VPS13B | |
| A 17 amino acid synthetic peptide near the amino terminus of human COH1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction