Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coilin Antibody (Pdelta) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB6001275
Description
Coilin Monoclonal specifically detects Coilin in Human, Mouse (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Microarray.Specifications
Coilin | |
Monoclonal | |
1.5 mg/mL | |
Western Blot 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Microarray | |
CLN80p80-coilin, coilin, coilin p80, p80 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Coilin (Pdelta) | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Pdelta | |
Unconjugated | |
P38432 | |
COIL | |
Recombinant fragment (C-terminus of Human coilin, 389 amino acids). | |
0.05 mL | |
Neuroscience | |
8161 | |
Human, Mouse (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction