Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Coilin Recombinant Rabbit Monoclonal Antibody (7Z4K1)
GREENER_CHOICE

Catalog No. PIMA535373
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535373 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535373 Supplier Invitrogen™ Supplier No. MA535373
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MAASETVRLR LQFDYPPPAT PHCTAFWLLV DLNRCRVVTD LISLIRQRFG FSSGAFLGLY LEGGLLPPAE SARLVRDNDC LRVKLEERGV AENSVVISNG.

The protein encoded by this gene is an integral component of Cajal bodies (also called coiled bodies). Cajal bodies are nuclear suborganelles of varying number and composition that are involved in the post-transcriptional modification of small nuclear and small nucleolar RNAs. The N-terminus of the coilin protein directs its self-oligomerization while the C-terminus influences the number of nuclear bodies assembled per cell. Differential methylation and phosphorylation of coilin likely influences its localization among nuclear bodies and the composition and assembly of Cajal bodies. This gene has pseudogenes on chromosome 4 and chromosome 14.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Coilin
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 7Z4K1
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene COIL
Gene Accession No. P38432, Q5SU73
Gene Alias C79982; CLN80; COIL; Coilin; coilin p80; hypothetical protein LOC507580; LOW QUALITY PROTEIN: coilin; nuclear phosphoprotein of Cajal body; p80; p80-coilin; wu:fb08h11
Gene Symbols COIL
Host Species Rabbit
Immunogen Synthetic peptide
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12812, 8161
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.