Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Coilin Monoclonal Antibody (Pdelta)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA125322
Description
Recommended positive controls: HeLa nuclear extract. MA1-25322 will not react with mouse. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is an integral component of Cajal bodies (also called coiled bodies). Cajal bodies are nuclear suborganelles of varying number and composition that are involved in the post-transcriptional modification of small nuclear and small nucleolar RNAs. The N-terminus of the coilin protein directs its self-oligomerization while the C-terminus influences the number of nuclear bodies assembled per cell. Differential methylation and phosphorylation of coilin likely influences its localization among nuclear bodies and the composition and assembly of Cajal bodies. This gene has pseudogenes on chromosome 4 and chromosome 14.
Specifications
Coilin | |
Monoclonal | |
1.9 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P38432 | |
COIL | |
Recombinant fragment corresponding to the 389 residue, C-terminal portion of human Coilin. | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot, Immunocytochemistry | |
Pdelta | |
Unconjugated | |
COIL | |
C79982; CLN80; COIL; Coilin; coilin p80; hypothetical protein LOC507580; LOW QUALITY PROTEIN: coilin; nuclear phosphoprotein of Cajal body; p80; p80-coilin; wu:fb08h11 | |
Mouse | |
Protein A | |
RUO | |
8161 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction