Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coilin Polyclonal Antibody
Coilin Polyclonal Antibody
Supplier: Thermo Scientific PA569975
Description
Coilin Polyclonal Antibody for Western Blot

Specifications
Coilin | |
Polyclonal | |
Unconjugated | |
COIL | |
C79982; CLN80; COIL; Coilin; coilin p80; hypothetical protein LOC507580; LOW QUALITY PROTEIN: coilin; nuclear phosphoprotein of Cajal body; p80; p80-coilin; wu:fb08h11 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
50998 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
COIL | |
synthetic peptide directed towards the C-terminal region of rat Coilin. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction