Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COL11A1 Polyclonal Antibody
COL11A1 Polyclonal Antibody
Supplier: Thermo Scientific PA566160
Description
COL11A1 Polyclonal Antibody for ICC/IF

Specifications
COL11A1 | |
Polyclonal | |
Unconjugated | |
COL11A1 | |
a1(XI) collagen; C530001D20Rik; cho; chondrodysplasia; CO11A1; COBA1; COL11A1; COLL6; Collagen alpha 1; collagen alpha-1(XI) chain; collagen type XI alpha 1 chain; collagen XI, alpha-1 polypeptide; collagen, type XI, alpha 1; pro-alpha1(XI) collagen; procollagen, type XI, alpha 1; STL2; STL3; XI chain precursor | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1301 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P12107 | |
COL11A1 | |
Recombinant Human COL11A1. Recombinant protein control fragment (Product #RP-106845). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction